Learn More
Research Products International Corp Vertical High Speed Tube Holder for 50mL Tubes, 3 Tube Capacity
Supplier: Research Products International Corp 155326
Tube Holder holds three 50mL tubes vertically. Holder snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse. Optimized for high speed vortexing a small number of tubes. No additional attachment necessary.