Learn More
TSI Incorporated 9535-A VelociCalc Air Velocity Meter
Supplier:  TSI Incorporated 9535A
Encompass
This meter measures and data logs several ventilation parameters using a single probe with multiple sensors. It measures velocity and temperature; calculates volumetric flow and data logs over 12,700 samples.
Catalog No. NC0644744