Learn More
BD VDRL Flocculation Antigens
Syphilis serology reagents
$216.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
B40764
|
BD
240764 |
5 mL |
Each for $216.00
|
|
|||||
B40765
|
BD
240765 |
0.5 mL |
Each for $271.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Nontreponemal test for syphilis
- For qualitative and semiquantitative determinations
- Suitable for testing cerebal spinal fluid (CSF) specimens
Specifications
VDRL Flocculation Antigen |