Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Walter Products™ Variable Volume Single Channel Micropipettes
With air displacement principle, tip ejector and disposable tips, this pipette is easy to operate for sampling and dispensing accurate liquid volumes.
Supplier: Walter Products™ TP20H
Description
- Large central pipetting button and separate ejection function
- 4-position volume display, which is always clearly visible
- Subjected to sterilization in the autoclave at 121°C (20 min), volume-change protection
- Easy calibration technique readjustment without special tools
- Short stroke of only 12.5 mm to reduce the risk of RSI (Repetitive Strain Injury)
Specifications
±0.8% | |
1 | |
Micropipette | |
0.02 μL |
Autoclavable | |
Sterile | |
2 to 20 μL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction