Learn More
Walter Products™ Variable Volume 12 Channel Micropipettes
SCI_ED
These multichannel micropipettes are lightweight and convenient to operate, requiring little pipetting strength.
Supplier:  Walter Products™ TP12D100H
 View more versions of this product
Catalog No. S139022
Add to Cart
Description
Description
  • High chemical and UV resistance
  • Completely autoclavable at 121°C
  • 4-position volume settings and a volume display which can easily be read
  • True one-hand operation for both right and left handed operators
  • Rotating dispensing head for optimal pipetting convenience
  • Due to optimized design on the tip cone, compatible with a wide range of tips
Specifications
Specifications
±0.8/1.4/4%
200/300 μL Tip
Micropipette
200/300 μL Tips
Autoclavable
12
10 to 100 μL
0.1 μL
Product Suggestions
Product Suggestions