Learn More
Electron Microscopy Sciences Variable Tilt Mount holder For Pin, 3.2 mm x 8mm Pin
Supplier: Electron Microscopy Sciences 7595205
This mount which is made from aluminum is used for the mounting and tilting of samples from 0 – 90 degrees as well as for SEM investigation with small working distances. The mount is marked for 30, 45, 70 and 90 degress. The table measures 11 x 11 mm (0.46" x0.46")and the overall size is 14 x 14 x12.7 mm
Type: 3.2 mm x 8mm Pin