Learn More
Mopec™ Variable Speed Knife Sharpener
This variable speed sharpener creates a precise angle and a consistent, exact bevel by sharpening, thinning, and honing which results in a stronger cutting edge.
Supplier: Mopec™ AH033
Description
- Honing wheels are mounted on parallel shafts that rotate in opposite directions
- Stainless steel case
- Replacement honing wheels available
- 220 V
Specifications
Knife Sharpener | |
Medium | |
Reusable | |
No |