Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit and Six Ports 1/16 in.
GREENER_CHOICE

Catalog No. 19830390
Click to view available options
Capacity (Metric):
13.5 L
20 L
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use With: Safe transport of hazardous liquids; Closure Material: Polypropylene; Color: White

  • For packaging group II and hazardous chemicals
  • Carbon filter prevents volatile organic compound vapors
  • Two-piece cap design

Specifications

Capacity (English) 3.6 gal.
Capacity (Metric) 13.5 L
Certifications/Compliance UN/DOT
Closure Material Polypropylene
Closure Size 70 mm
Color White
Description Six (6) 1/16 in. Tubing OD Ports
Dimensions (L x W x H) 287 x 235 x 333 mm
For Use With (Application) Safe transport of hazardous liquids
Height (English) 13.1 in.
Length (English) 11.3 in.
Material HDPE
Product Type UN/DOT Carboy with Filter Kit
Shape Rectangular
Width (English) 9.25 in.
Show More Show Less
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.