Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ VaporTrap™ Polypropylene, 83mm Carboy Caps with Filter Kit, Four Ports 1/8” OD Tubing, Three Ports 1/4 in. OD Tubing
Click to view available options
Description:
Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
Description
- Carbon filter prevents volatile organic compound vapors
Specifications
Product Type | Cap |
Description | Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports |
Dimensions (L x W x H) | 5 x 5.5 x 11 in. |
Material | Polypropylene |
For Use With (Equipment) | carboys with 83mm cap |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction