Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
VapLock™ Solvent Waste Kits for Bottles and Carboys collect effluent from multiple sources while minimizing harmful solvent exhaust
Supplier: Cole-Parmer™ 1201905
Description
Various cap sizes available to fit your solvent containerWarranty and Services
None
Specifications
Solvent Safety | |
PTFE | |
Solvent Waste Kit |
Solvent-Compatible Containers | |
DIN 60 | |
1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F) |