Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps with Inlet Valve and Filter
VapLock™ Solvent Delivery Caps with Inlet Valve and Filter reduce chemical evaporation and exposure to hazardous vapors
Supplier: Cole-Parmer™ 1201846
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) ODWarranty and Services
None
Specifications
Solvent-Compatible Containers | |
Solvent Safety | |
White PTFE, PP, SS, Viton |
Wako 38 mm | |
Two 1/4 in.-28 ports | |
1/4 in.-28 UNF(F) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction