Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps with Inlet Valve and Filter
VapLock™ Solvent Delivery Caps with Inlet Valve and Filter reduce chemical evaporation and exposure to hazardous vapors
Supplier: Cole-Parmer™ 1201832
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) ODWarranty and Services
None
Specifications
Solvent Safety | |
1/4 in.-28 UNF(F) | |
White PTFE, PP, SS, FFKM |
Two 1/4 in.-28 ports | |
Solvent-Compatible Containers | |
Nalgene 38-430 |