Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps with Inlet Valve and Filter
VapLock™ Solvent Delivery Caps with Inlet Valve and Filter reduce chemical evaporation and exposure to hazardous vapors
Supplier:  Cole-Parmer™ 1201830
 View more versions of this product
Catalog No. 13-311-030
Add to Cart
Description
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) OD
Warranty and Services
None
Specifications
Specifications
Solvent-Compatible Containers
White PTFE, PP, SS, EPDM
Nalgene 38-430
Solvent Safety
Two 1/4 in.-28 ports
1/4 in.-28 UNF(F)
Product Suggestions
Product Suggestions