Learn More
Cole-Parmer Davis Instruments 6152 Wireless Weather Station
Supplier: Cole-Parmer 8640303
This powerful station transmits data up to 1000 ft or even further with an optional wireless repeater. Measured parameters update every 2.5 seconds as compared to 3 to 15 minutes found on other stations.