Learn More
Apexbio Technology LLC Vanillic acid,100mg CAS# 121-34-6
Supplier: Apexbio Technology LLC B7917100
Vanillic acid is used as a precursor in the synthesis of a wide range of bio-based epoxy resins and polyesters.It is also a key starting material in the total synthesis of barmumycin4 and bosutinib. Other sizes are also available. Please inqury us for quote.
This item is not returnable.
View return policy