Learn More
RADWELL INTERNATIONAL VALVE, PTFE DIAPHRAGM, NORMALL
Supplier:  RADWELL INTERNATIONAL BSDA025TNCTF
Encompass_Preferred
VALVE, PTFE DIAPHRAGM, NORMALLY CLOSED,AIR TO OPEN PROCESS, CONNECTION SIZE1/4",CONNECTION TYPE THREADED,MAXPRESSURE AT 40 - 75 DEGREES F 6.8PSI, AIR INLET CONNECTION 1/8", FAIL DRY
Catalog No. NC2166614