Learn More
Perkin Elmer US LLC Valve O-Ring
Supplier: Perkin Elmer US LLC 02401293
To keep your Elemental Analyzer operating efficiently, you should clean and rebuild the Normally Closed Valve periodically. Maintenance instructions are included in your operator's manual. Maintenance of the Normally Open Valve requires a PerkinElmer Engineer.