Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Purple
Click to view available options
Size:
X-Small
Small
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
- Fluid resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Hip length
- Autoclavable and washable up to five times
- Latex free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Size | Small |
Gender | Unisex |
Material | Polypropylene |
Color | Purple |
Latex Free | Yes |
Model | 3630PP |
No. of Pockets | 3 |