Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ ValueWare™ Wide-Mouth Erlenmeyer Flasks
SDPSCI_ED

With capacity scale.

Supplier:  DWK Life Sciences 566501000

 View more versions of this product

Catalog No. S00153



Add to Cart

Description

Description

  • Heavy-duty Kimax KG-33 glass flasks
  • Tooled top finish

Compliance

Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E1404, Type I, Class 2 specification requirements.

Specifications

Specifications

Rubber
4.8 in.
Borosilicate Glass
50 mL
21.5 cm
12.3 cm
White Ceramic Enamel scale graduations; Top style: Reinforced and tooled with rounded finish beaded
1000 mL
250 to 1000 mL
8.46 in.
Wide
11
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit