Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ ValueWare™ Wide-Mouth Erlenmeyer Flasks
SDPSCI_ED

With capacity scale.

Supplier:  DWK Life Sciences 56650250

 View more versions of this product

Catalog No. S00151

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

  • Heavy-duty Kimax KG-33 glass flasks
  • Tooled top finish

Compliance

Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E1404, Type I, Class 2 specification requirements.

Specifications

Specifications

Rubber
3.07 in.
Borosilicate Glass
25 mL
13 cm
78 mm
White Ceramic Enamel scale graduations; Top style: Reinforced and tooled with rounded finish beaded
250 mL
50 to 225 mL
5.11 in.
Wide
8
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit