Learn More
DWK Life Sciences Kimble™ ValueWare™ Round-Bottom Boiling Flasks
Made from Kimax™ Type KG-33 glass.
Supplier: DWK Life Sciences 55285500/EMD
This item is not returnable.
View return policy
Description
- Short neck
- Full-length standard taper 24/40 joint
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E1403, Type II, Class 3 specification requirements.
Specifications
4 in. | |
Glass | |
16 cm | |
24/40 | |
102 mm |
500 mL | |
6.2 in. | |
Short | |
Round |