Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI DECON-QUAT 200C Fifth Generation Quaternary Ammonium Solution, Sterile
DECON-QUAT 200C Fifth Generation Quaternary Ammonium Solution is formulated to disinfect, clean, and sanitize hard, non-porous surfaces.
Supplier: VAI DQ200C038Z
This item is not returnable.
View return policy
Description
- Filled in ISO 5 (Grade A/B, Former Class 100)
- Filtered at 0.2 microns
- Gamma irradiated to a sterility assurance level of 10-6
- Sterility tested according to current USP compendium
- Completely traceable from start to finish
- Completely validated for sterility and shelf life
Specifications
Quaternary Ammonium | |
Bottle | |
8 oz. |
Sterile | |
Concentrate | |
237 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction