Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Valsartan
High affinity, selective AT1 antagonist
$237.80 - $989.48
Specifications
Purity | >99% |
---|---|
Gene Alias | CGP 48933 |
Product Type | Valsartan |
Molecular Weight (g/mol) | 435.52 |
Inhibitors | Valsartan |
Description
High affinity AT1 receptor antagonist (Ki = 2.38 nM). Displays 30,000-fold selectivity over AT2 receptors. Inhibits angiotensin II-induced release of aldosterone in vitro. Orally active.Specifications
>99% | |
Valsartan | |
Valsartan |
CGP 48933 | |
435.52 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title