Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valproic Acid, USP, 98-102%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
Poly-Coated Glass Bottle
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
2-propylpentanoic acid
CCCC(CCC)C(O)=O
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
2-propylpentanoic acid
CCCC(CCC)C(O)=O

V2011, 99-66-1, C8H16O2

Spectrum™ Chemical Valproic Acid, USP is used to treat certain types of seizures. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Ignition Residue 0.001
Packaging Poly Pail
Quantity 5 kg
Percent Purity 98 to 102%
Grade USP