Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI WipeDown Dry Wipes
Ultra-clean laundered and suitable for use in cleanroom environments. WipeDown Dry Wipes, for use in ISO 4, are 12 x 12 in. size and constructed of 100% continuous filament polyester designed to be clean and absorbent.
Supplier: VAI VEL812X12S3002
This item is not returnable.
View return policy
Description
- For use during wipe downs of filling and packaging machinery, stainless steel surfaces, Lexan, polycarbonate, glass, and any other critical surface that requires cleaning
- Laser cut sealed edge
- Size: 12 x 1 2 in.
Specifications
Dry Wipe | |
Laser cut sealed | |
Wipe downs of filling and packaging machinery, stainless steel surfaces, Lexan, polycarbonate, glass, and any other critical surface cleaning |
Polyester fiber | |
30.48 x 30.48 cm (12 x 12 in.) | |
Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title