Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WFI Quality Water, VAI
Click to view available options
Quantity:
1 gal. (3.8 L)
16 oz. (473 mL)
2 gal. (7.6 L)
325 mL (11 oz.)
Description
- Filled in an ISO 5 (Grade A/B, Former Class 100) operation
- Filtered at 0.2 microns
- Components are air washed with 0.2 micron filtered air before assembly
- Lot sterility tested according to current USP compendium
- Lot tested for endotoxins
Specifications
Format | Liquid |
Quantity | 16 oz. (473 mL) |
Volume (English) | 16 oz. |
Safety and Handling
ShelfLife | 2 Years |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction