Learn More
USP Purified Steri-Water, VAI
USP Purified Steri-Water designed for use in the pharmaceutical, biotechnology, and medical device industries for chemical formulations, cleaning, rinsing and many other applications.
Supplier: VAI STWA2G
Description
- Specifically formulated as a sterile pharmaceutical cleanroom formula
- Lubricates for continuous and effortless manufacturing
- Designed for all washable environmental surfaces
- Filled in an ISO 5 (Grade A/B, Former Class 100) operation
- Filtered at 0.2 microns
- Components are air washed with 0.2 micron filtered air before assembly
Specifications
Sterile | |
Filtered | |
2 gal. |
Lot specific Certificate of Analysis, Certificate of Sterility, and Certificate of Irradiation | |
Bottle | |
7.6 L |
Safety and Handling
ShelfLife : 2 Years