Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI STERI-PEROX™ Non-sterile Hydrogen Peroxide Solution
STERI-PEROX™ Non-sterile Hydrogen Peroxide Solution is designed for pharmaceutical and biotechnology operations.
Supplier: VAI SPER016%
Includes: Lot-specific Certificate of Irradiation and Certificate of Analysis
Description
- Ideal for use on walls, ceilings, floors and surfaces
- Formulated with USP purified water and filtered at 0.2 μm
- Completely traceable and validated for sterility and shelf life
Order Info
Double-bag packaging.
Compliance
Completely tested according to current USP compendium.
Specifications
Non-sterile | |
6% | |
Sterilization of manufacturing, packaging and filling equipment in aseptic processes, for use in cleaning rotation cycles to maintain a clean environment |
USP, ISO | |
Filtered |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title