Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI Process2Clean 6 Chlorinated Alkaline Cleaner
Process2Clean 6 is a chlorinated alkaline cleaning agent. P2C 6 contains buffering agents to maintain substrate integrity. P2C 6 is capable of removing highly proteinaceous soils, oils, waxes, creams, and polymers.
$333.00 - $3486.00
Specifications
Form | Liquid |
---|---|
pH | 12.6 |
Appearance | Clear |
Content And Storage | 89.6°C to 176°C |
For Use With (Application) | For low energy agitate immersion systems, pressure spray, or manual washing |
Catalog Number | Mfr. No. | Volume (Metric) | Volume (English) | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Volume (English) | Sterility | Price | Quantity & Availability | |||
19-546-906
|
VAI
PC61G01 |
3.8 L | 1 gal. | Non-sterile |
Case of 4 for $344.00
|
|
|||
19-546-907
|
VAI
PC61G02 |
3.8 L | 1 gal. | Sterile |
Case of 4 for $469.00
|
|
|||
19-546-908
|
VAI
PC630G01 |
113.6 L | 30 gal. | Non-sterile |
Each for $1,324.00
|
|
|||
19-546-909
|
VAI
PC630G02 |
113.6 L | 30 gal. | Sterile |
Each for $2,274.00
|
|
|||
19-546-910
|
VAI
PC655G01 |
208.2 L | 55 gal. | Non-sterile |
Each for $2,089.00
|
|
|||
19-546-912
|
VAI
PC65G01 |
18.9 L | 5 gal. | Non-sterile |
Each for $333.00
|
|
|||
19-546-911
|
VAI
PC655G02 |
208.2 L | 55 gal. | Sterile |
Each for $3,486.00
|
|
|||
19-546-913
|
VAI
PC65G02 |
18.9 L | 5 gal. | Sterile |
Each for $457.00
|
|
|||
Description
- Characteristic: Alkaline
- Clarity: Clear
- Appearance: Yellow
- pH (1% solution) : 11.0 – 13.0
- Conductivity (1% solution): 6.50 – 8.50 mS
Specifications
Liquid | |
Clear | |
For low energy agitate immersion systems, pressure spray, or manual washing |
12.6 | |
89.6°C to 176°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title