Learn More
Sklar Instruments Pederson Vaginal Speculum, Sklar 6-1/2 in., 6-1/2 in. x 1-1/2 in., Blunt, Straight, Extra Long, Stainless Steel. EA.
Supplier: Sklar Instruments 98366
Pederson Vaginal Speculum, Sklar. 6-1/2 in., 6-1/2 in. x 1-1/2 in., Blunt, Straight, Extra Long, Stainless Steel. The Merit Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. This product is an extra-long sized speculum with 6-1/2 in. X 1-1/2 in. blades.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.