Learn More
Chemglass Life Sciences Vacuum Trap, O-Ring Style, Stopper Only, #30 O-Ring Joint
SDP
Supplier:  Chemglass Life Sciences CG451330
Encompass_Preferred
Vacuum Trap, O-Ring Style, Stopper Only, #30 O-Ring Joint. Similar to CG-4515 traps but having an o-ring joint to connect the trap body to the stopper. Serrated hose connections are 13.5mm at the smallest serration and 15.5mm at the largest serration. STOPPER ONLY! Pictured is the complete vacuum trap assembly.
Catalog No. 50-153-1741
Explore available promotions