Learn More
Chemglass Life Sciences Vacuum System, Self-Adjusting, PC3004 Vario Select, Touch Screen Controller, 100-120V
SDP
Supplier:  Chemglass Life Sciences CG4802104
Encompass_Preferred
Request Bulk Order
Vacuum System, Self-Adjusting, PC3004 Vario Select, Touch Screen Controller, 100-120V. The PC3004 VARIO select is the best choice vacuum system for bench-top sized rotary evaporators and other evaporative applications. The PC3004 VARIO select controls the vacuum in your rotary evaporator with minimal supervision, continuously optimizing to changing vapor conditions without programming. Just press "start" and the PC3004 VARIO select provides faster evaporation and tighter control, while freeing you up to do more important work. It features a VARIO version of our MD1C vacuum pump, with integrated solvent recovery in a compact, attractive package.Touch Screen Controller:The integrated VACUU-SELECT controller comes with automatic operating modes for most common lab vacuum applications. Drag and drop menus let you set up your own custom operations in minutes.
Catalog No. 50-197-9518
Explore available promotions