Learn More
Cole-Parmer Cole-Parmer Air diaphragm vacuum/pressure pump, 0.37 cfm, 115 VAC
Supplier: Cole-Parmer 7920200
This portable vacuum pump is designed for versatility with a tough, lightweight plastic housing and integral carrying handle. The long-wearing motor delivers performance characteristics suitable for a wide range of applications. Pump features a convenient cord storage compartment in its base. Note: Do not pump liquids or immerse pump. Avoid organic, basic, and acidic vapors.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.