Learn More
Cole-Parmer Gardner Denver PTFE-Coated Vacuum Pump, Gauge/Reg/Valve; 0.75 cfm/23.2"Hg-25psi/115V
Supplier: Cole-Parmer 7920000
Pumps include suction cup feet and a 6-ft power cord with built-in switch (the 115 VAC models have grounded plug, 220 VAC and DC models have stripped ends).