Learn More
Electron Microscopy Sciences Vacuum Oven Model 3608-5, 120 V, 10"W x 10"H x 12"D chamber, 0.7 cu. ft (19.8 L) Capacity
Supplier:  Electron Microscopy Sciences 6323610
Encompass_Preferred
Multipurpose vacuum oven with 220 deree C maximum temperature conserves chamber space for drying, curing, vacuum embedding, and plating applications. Silicone door gasket and a positive latch door maintains the seal for all vacuum levels. Exclusive Polycarbonate safety shield protects door glass. Two removable aluminum shelves including standard. Easy-to-clean #304 staniless steel chamber interior. Powder coated heavy guage steel exterior construction.
Chamber: 10"W x 10"H x 12"D
Capacity: 0.7 cu. ft (19.8 L)
Thermometer: LED
Catalog No. 50-313-12