Learn More
Integra Biosciences Corp VACUBOY HANDOPERATOR SET
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Integra Biosciences Corp 155500
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
VACUBOY hand operator-Set consists of: - VACUBOY hand operator - Stand for hand operator - Rubber adapter for Pasteur Pipettes - 1-channel stainless steel aspiration adapter 40 mm - 8-channel stainless steel aspiration adapter 40 mm - 1-channel plastic aspiration adapter for disposable tips - 1-channel plastic aspiration adapter for disposable tips with ejector - 8-channel plastic aspiration adapter for disposable tips with ejector $ 360.00 $ 342.00 $ 324.00 155510 VACUBOY hand operator with 1-Channel stainless aspiration adapter 40 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.