Learn More
Foxx Life Sciences VactrapXL™ Vacuum Trap System (Vacuum Flask) for Aspiration and Vacuum Protection, Shatterproof 4+4 Liter Aspirator Bottle Assembly with Secondary Container, Autoclavable Polypropylene (PP), 4L Bottle with 4L Overflow Bottle and Inline Vent Filter
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Foxx Life Sciences 3054401FLS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Foxx Life Sciences VactrapXL™ is the world's most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, VactrapXL safely contains waste from supernatural removal, chemical separation, and cell or tissue culture media aspiration. Best of all, VactrapXL™ comes ready to use; simply attach to your vacuum and begin your work. The full VactrapXL™ system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap® closure for tangle-free opening, no need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.