Learn More
Foxx Life Sciences VACTRAP ASPIRATOR HDPE 1L BIN
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Foxx Life Sciences 3021001FLS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Foxx Life Sciences' Vactrap2™ is the world's most advanced vacuum trap system. Ideal for the protection of vacuum lines and scientists alike, Vactrap2™ safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap2™ comes ready to use; simply attach to your vacuum and begin your work. The full Vactrap2™ system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap closure for tangle-free opening, no need to disconnect tubing, ever. NEW Improved heavy-duty EZBio TPE V tubing to avoid kinking. A 0.2µm PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nestled neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for the modulation of vacuum. Other Vactrap2™ options offer one bottle or setups without secondary containment.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.