Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-FORM™ Safety Harness - Qwik-Fit Leg Straps

The V-FORM™ full body safety harness allows you to focus on your job - not your harness

Supplier:  MSA™ 10197198

 View more versions of this product

Catalog No. 19-001-668


Add to Cart

Description

Description

  • Featuring a patent-pending RaceFORM™ buckle, bulky chest straps are eliminated for a close and comfortable fit.
  • The athletic cut of the V-FORM harness contours to the body improving upper torso movement on the job
  • Pull-down adjustment allows for quick and easy harness adjustment that ensures a proper fit that lasts throughout the work day
  • Variety of configurations available
Specifications

Specifications

Quick-Connect
400 lb.
ANSI Z359.11-2014 CSA Z259.10
Polyester
66 cm
Harness
181 kg
Back
RaceFLEX™ Quick-Connect Buckle
Working at Heights
Vest
Fall Protection
2.5 lb.
2X-Large
26 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit