Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP Scientific Freestanding / Mobile Glassware Washers
Designed for ease of operation and versatile loading, SP Hotpack Freestanding / Mobile Laboratory Glassware Washers include sturdy casters that allow flexible placement in the laboratory. Equipped with premium features including a soft touch controller, enhanced drying, and quiet run cycles, SP Hotpack Laboratory Glassware Washers are engineered for durability and energy efficient operation.
Supplier: SP Scientific UCWHA30101
Description
- Provide reliable day-in, day-out performance so your laboratory glass is always ready-to-go
- All models come with removable top and bottom stainless-steel open racks to accommodate a variety of glassware
- Spindle rack ready models accommodate direct injection spindle racks (sold separately) for thorough cleaning inside narrow-neck glassware
- Stainless steel baskets, insert racks and accessories sold separately to maximize space, improve efficiency, and protect specialty glassware
- Temperature selection to 70°C (158°F); can effectively dissolve stubborn oils
- DI rinse models ensure a spotless finish; easily connect to fixed or portable DI water systems
- Optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18psi
- Delayed start feature enables washer to operate unattended during off-peak hours to reduce energy costs
- Designed for optimal quiet operation with noise buffering insulation
- Three filters protect pumps from debris; conveniently located for easy access
- Minimal, efficient water consumption
- Enhanced dual fan drying system for spotless results
- 304 stainless steel chamber and exterior casing ensures corrosion protection and long life
- 3 in. lockable casters allow easy, controlled mobility of unit
- UL 61010-1 and CSA 22.2 61010-1 certified by MET Labs
- Microprocessor control system features 6 cycle selections including a customizable timed option to ensure minimum energy consumption. All wash cycles have pre-wash, wash, pre-rinse and rinse cycles to prevent contamination.
- Rinse Only - Short tap water rinse cycle
- Normal Wash - Lower temperature (55°C) longer cycle with three (3) final rinse cycles
- Daily Wash - Standard temperature (65°C) with two (2) final rinse cycles
- Heavy Wash - Higher temperature (70°C) longer cycle with multiple pre-wash and final rinse cycles
- Heavy Wash Time Saver - Lower temperature (55°C) version of Heavy Wash with a shorter overall cycle time
- Timed Wash Programs - Standard temperature (65°C) in programmable 15 minute increments up to 2.5 hours
Specifications
DI Rinse and High Performance Pump - Top and Bottom Spindle Rack Ready | |
Stainless Steel | |
Min: Tap 2.4 gal. (9L), DI 0.8 gal. (3L); Max: Tap 4 gal. (15L), DI 1.6 gal. (6L) | |
Washing Lab Glassware | |
24.25 in. | |
38.5 in. | |
97.8 cm | |
24.0 in. | |
136.5 lb. | |
120V, 60Hz, 15A | |
60 Hz |
Digital | |
60 psi | |
MET Labs | |
55°-70°C (131°-158°F) | |
61.6 cm | |
25 in. | |
63.5 cm | |
61.0 cm | |
62 kg | |
120 V Plug in |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction