Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI V-100 Vacuum Pump
SDP

The Vacuum Pump V-100 is a chemically resistant PTFE diaphragm vacuum pump with space saving design. It covers essential needs in vacuum generation.

Supplier:  BUCHI 11V100111

 View more versions of this product

Catalog No. 05-405-207


Add to Cart

Description

Description

  • Effective vacuum source for limited budgets
  • Small footprint saves valuable laboratory space
  • Light weight and compact design with integrated carrying handle
  • High up time due to low maintenance
  • Accurate vacuum regulation thanks to optional interface
  • Maximum longevity due to: Robust and stable design and Integrated ECO2 mode
  • On/off-regulated by the Interface I-100, optimally suited to work with Rotavapor R-100
  • Includes silencer and 2m of vacuum tubing
Specifications

Specifications

10 mbar
Vacuum Pump and Controller
1.5 m3/hr.
IP 20
Atmospheric
I-100 Interface, Woulff Bottle, and Secondary Condenser with Insulation
V-100 and I-100
Vacuum System
Rotary evaporation
100/240 V
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit