Learn More
Honeywell™ Uvex™ Replacement Lenses for Flex Seal™ Goggles
Click to view available options
Lens Tint:
Gray
Lightweight, silicone body conforms to assure the highest level of comfort and eye protection. Exceptional chemical splash and impact resistance. Fits over most prescription eyewear.
Uvex Flex Seal goggle is made of a lightweight, silicone body, which conforms to the face, to assure the highest level of comfort and eye protection. The silicone body stays supple even in extreme temperatures and seals to almost any face shape, while offering exceptional all-day comfort.
  • Available in HydroShield or Uvextreme Anti-Fog lens coatings
  • Fits comfortably over prescription eyewear
  • Available in various lens tints
  • Economical lens replacement
Lens Tint Gray
Product Type Replacement lens
For Use With (Equipment) Flex Seal Goggles