Learn More
Honeywell™ Uvex™ Futura™ Chemical-Splash Goggles
Click to view available options
Large
Description
Includes
Neoprene headstrap.
- Slip easily over most prescription spectacles; adapt to respirators and hardhats without impeding fit
- Lightweight body molds to the face for a firm seal and comfortable protection
- Indirect venting protects eye area against hazardous splashes and airborne particles while allowing ample air circulation for comfort
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Lens Type | Uvextreme AF |
Certifications/Compliance | ANSI Z87.1-2010, CSA Z94.3 |
Lens Tint | Clear |
Product Type | Chemical Splash Protection Goggles |
For Use With (Equipment) | Fits over most glasses |
Includes | Neoprene headband |
Size | Large |
Material | Polycarbonate |