Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Uvex Flex Seal™ Safety Goggles
Click to view available options
Headband Material:
Fabric
Neoprene
Description
- Lightweight, silicone body conforms to the face assuring the highest level of comfort and eye protection
- Hydroshield™ AF coating provides 2x greater scratch resistance than other Honeywell™ anti-fog coatings
- Uvextreme AF lens coating offers consistent fog-free time 90x longer than the majority of other anti-fog products
- Tested by Honeywell™ to meet the requirements of the CSA Z94.3 standard
- Silicone body stays supple even in extreme temperatures
- Exceptional chemical splash and impact resistance
- Fits over most prescription eyewear
- Indirect venting system
- Made in the USA
Specifications
Lens Coating | Anti-fog |
Certifications/Compliance | ANSI Z87+ (High Impact), ANSI Z.87.1/1989, CA 21,199, CSA Z94.3 |
Lens Tint | Clear |
Product Type | Safety Eyewear |
For Use With (Application) | Abrasions, Airborne Particulate, Biohazard, Chemical, Impact, Splash |
Frame Color | Navy |
Headband Material | Fabric |
Includes | Headband, Anti-Fog lens |
No. per Case | 50/Pk. |
No. per Pack | 1 Ea. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction