Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Uvex Clear™ Plus Lens Cleaner Solution
Click to view available options
Quantity: 16 oz.
Description
- Swiftly lifts and removes dirt, dust, debris, oil
- Reduces visible soil to below detectable limits (.2 to .3% haze)
- Streak-free, residue-free, haze-free results
- Alcohol and silicone-free
- Methanol, ethanol, and isopropanol-free
- Butoxyethanol-free
- Pleasant smell from all natural papaya extract
- Safe to use on any lens or coated surface
- Won’t degrade or delaminate lens material
- Can be used in temps as low as -40°C
Specifications
Product Type | Cleaning solution |
For Use With (Application) | Uvex Clear™ Plus Lens Cleaner products |
Quantity | 16 oz. |
Material | Lens Cleaner Solution (Liquid) |