Learn More
Honeywell™ Uvex™ Astro OTG 3001™ Safety Eyewear
Click to view available options
Clear lens; Blue frame; Uvextreme anti-fog coating
Specifications
Lens Coating | Anti-fog |
Certifications/Compliance | ANSI, CSA |
Lens Tint | Clear |
Product Type | Safety Eyewear |
Frame Color | Blue |
Weight (Metric) | 50 g |
Lens Material | Polycarbonate |
Frame Material | Nylon |