Learn More
Honeywell™ Uvex™ Ambient OTG Safety Eyewear Replacement Lens
Click to view available options
Replacement lens; Lens tint: shade 5.0; Lens coating: ultra dura
Description
- The Uvex™ Ambient provides over-the-glass coverage, offering your workers full protection while wearing their own prescription eyewear
- Available with a range of comfort features and in two sizes, the Uvex™ Ambient OTG offers all-day coverage for a variety of workers and a wide range of prescription eyewear styles
- This high performance model can improve your workers' productivity as well as lower your cost-of-ownership with Uvex's renowned tints and lens replacement system
- The Uvex™ Ambient™ OTG is lightweight, comfortable and a stylish alternative to prescription safety eyewear
- Exceptional peripheral vision and coverage in a lightweight, wraparound uni-lens design
- Economical and easy lens replacement allows multiple tints and extends the life of the eyewear
Compliance
Meet ANSI™ Z87.1-2003. Certified to CSA Z94.3 standards.
Specifications
Certifications/Compliance | ANSI Z87.1-2003 and CSA Z94.3 standards |
Lens Tint | IR Shade 5 |
Product Type | Replacement Lens; Over-the-glasses |
For Use With (Equipment) | For safety eyewear |
For Use With (Application) | Cutting operations |