Learn More
Hach Company Replacement UV Lamp for Millipore Water Systems
Supplier: Hach Company 2512700
Used for the destruction of organic contaminants in water by photo-oxidation. 185nm.Note: Cannot be sold into CT, LA, ME, RI and VT.