Learn More
Sklar Instruments Sims Uterine Sound, Sklar 12-1/2 in., Blunt, Curved, Smooth, Solid Flat, Silver Plated Stainless Steel. EA.
Supplier: Sklar Instruments 98384
Sims Uterine Sound, Sklar. 12-1/2 in., Blunt, Curved, Smooth, Solid Flat, Silver Plated Stainless Steel. The Merit Sims Uterine Sound is a malleable, silver plated OB/GYN instrument used for probing and dilating the uterus as well as measuring the length of the cervical canal. It is very similar to the Simpson uterine sound. The instrument is available with or without graduated centimeter markings. This product is straight, but malleable and can be curved as necessary. The handle is a short, smooth thumb grip that features 3 vertical grooves for improved grip. It does not have any measurement markings. Overall, the length is 12-1/2 inches.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.