Learn More
Instech USPHS0100.5NJ1P-50 SOLUTION
Supplier: Instech USPHS0100.5NJ1P50
Pharmaceutical-grade catheter flush solution prepared with heparin sodium following the USP <797> standard for compounding sterile preparations. The solution contains heparin (10 USP units/ml) in 0.9% saline, 50 pcs (10 packs of 5) 0.5 ml in NORM-JECT-F 1ml slip fit syringe, PinPort injector